EDTA, pH 8.0, 0.5M
EDTA, pH 8.0, 0.5M
0.5M EDTA, 20 L, Cubitainer®
Catalog # 75800-474
EDTA, pH 8.0, 0.5M
Catalog # 75800-474
Supplier:  
CAS Number:  
EDTA, pH 8.0, 0.5M
0.5M EDTA, 20 L, Cubitainer®
CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
SOLUTION EDTA RNA STORAGE 0.5M 1 X 10L Catalog #75800-472
Product 75800-472 is a replacement for 75800-474 but varies by Brand, Packaging or Quantity.
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ethylenediaminetetraacetic acid is a polyamino carboxylic acid, commonly used to deactivate metal dependent enzymes, or as a chelating agent that binds to metal ions
Suitable for RNA storage. Offered in different molar concentration for all common biological applications.
Spécifications
Spécifications
  • Type de conditionnement:
    Cubitainer®
  • Désignation:
    0.5M EDTA
  • Taille:
    20 L