Biox Transfer Pipettes
160 mm, Sterile, To 3 mL, Individually Wrapped
Catalog # 75799-908
Supplier: TWD Scientific
Fournisseur #: BX30-0138A1-1
Jump to:
Product Details & Documents
Transfer Pipettes are perfect for quick and safe liquid transfer.
- Disposable
- Polyethylene construction
- Uniform drop size
- 100% inert for accurate sampling
Polyethylene pipettes are a great alternative to pasteur pipettes or rubber bulbs.
Spécifications
- Sterility:Sterile
- Longueur:160 mm
- Packed:Individually Wrapped
- Graduation:To 3 mL