Biox Cell Spreaders
Orange, Individually Wrapped
Catalog # 75799-958
Supplier: TWD Scientific
: BX65-1001
Jump to:
Product Details & Documents
Cell Spreaders are ideal for the even spreading of bacterial cultures.
- Polystyrene construction
- L-Shape design
- Disposable
- Smooth spreading surface with slight upward angle reduces the risk of damaging agar surfaces
- Gamma radiation sterilized
- Individually wrapped
L-Shaped cell spreaders allows for application on the entire culture dish or plate surface.
Spécifications
- Color:Orange
- Packaging:Individually Wrapped