Repeater® E3 and E3x Pipettors, Eppendorf® | Avantor
Repeater® E3 and E3x Pipettors, Eppendorf®
Repeater® E3x Bundle, Including Charger Stand
Catalog # 75796-402
Fournisseur #:  4987000410
Repeater® E3 and E3x Pipettors, Eppendorf®
Catalog # 75796-402
Supplier:  
Repeater® E3 and E3x Pipettors, Eppendorf®
Repeater® E3 and E3x Pipettors, Eppendorf®
Repeater® E3x Bundle, Including Charger Stand
Discontinued
Product Details & Documents
The Repeater® E3/E3x Pipettors, in combination with the Combitips Advanced® pipette tips, creates a versatile dispensing system that functions according to the positive displacement principle.
  • Automatic Combitip® recognition
  • Ergonomic one-button tip ejector
  • Precisely programmable aspiration and dispensing speeds
  • Repetitive dispensing modes
  • Wide dispensing range: 1 μl to 50 ml, in 100-nanoliter increments
  • Safe and ergonomic operation with 9 different selectable languages
These devices provide precise and accurate dispensing of viscous solutions (e.g. glycerol) or liquids with high vapor pressure (e.g. ethanol). Contamination-free dispensing for toxic, radioactive, and other hazardous liquids (all available modes). Large illuminated color display reduces eye strain when reading. Motor-driven function reduces human error to a minimum, and reduces the risk of repetitive strain injuries (RSI).
It is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results.
Spécifications
  • Désignation:
    Repeater® E3x Bundle, Including Charger Stand