WaterPro® Bladder Tanks, Labconco | Avantor
WaterPro® Bladder Tanks, Labconco
WaterPro® RO Bladder Tank, 20 Gal. (75.7 L), for WaterPro® RO System
Catalog # 75796-358
Supplier:   Labconco
Fournisseur #:  9201000
WaterPro® Bladder Tanks, Labconco
Catalog # 75796-358
Supplier:  Labconco
WaterPro® Bladder Tanks, Labconco
WaterPro® Bladder Tanks, Labconco
WaterPro® RO Bladder Tank, 20 Gal. (75.7 L), for WaterPro® RO System
Price
$2,730.00
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
WaterPro® RO Bladder Tanks provide additional storage of reverse osmosis (RO) water produced by the WaterPro® RO System (sold separately)
The pressurized water can be distributed to a polishing system or glassware washer through either of two pressurized ports. Includes all tubing and hardware for connection. Available in 14 or 20 gallon sizes.
Spécifications
  • Désignation:
    WaterPro® RO Bladder Tank, 20 Gal. (75.7 L), for WaterPro® RO System