WaterPro® Bladder Tanks, Labconco
WaterPro® RO Bladder Tank, 20 Gal. (75.7 L), for WaterPro® RO System
Catalog # 75796-358
Supplier: Labconco
Fournisseur #: 9201000
Jump to:
Product Details & Documents
WaterPro® RO Bladder Tanks provide additional storage of reverse osmosis (RO) water produced by the WaterPro® RO System (sold separately)
The pressurized water can be distributed to a polishing system or glassware washer through either of two pressurized ports. Includes all tubing and hardware for connection. Available in 14 or 20 gallon sizes.
Spécifications
- Désignation:WaterPro® RO Bladder Tank, 20 Gal. (75.7 L), for WaterPro® RO System