WaterPro® Bladder Tanks, Labconco
WaterPro® RO Bladder Tank, 14 Gal. (53 L), for WaterPro® RO System
  75796-356
 :   Labconco
 :  9201001
WaterPro® Bladder Tanks, Labconco
  75796-356
 :  Labconco
WaterPro® Bladder Tanks, Labconco
WaterPro® Bladder Tanks, Labconco
WaterPro® RO Bladder Tank, 14 Gal. (53 L), for WaterPro® RO System
$2,480.00
Each
WaterPro® RO Bladder Tanks provide additional storage of reverse osmosis (RO) water produced by the WaterPro® RO System (sold separately)
The pressurized water can be distributed to a polishing system or glassware washer through either of two pressurized ports. Includes all tubing and hardware for connection. Available in 14 or 20 gallon sizes.
  • Désignation:
    WaterPro® RO Bladder Tank, 14 Gal. (53 L), for WaterPro® RO System