Thumb Dressing Forceps, Electron Microscopy Sciences
Product Details & Documents
These conventional forceps are very popular and are available in a variety of length, tip, and handle styles.
- Made of premium grade, hardened stainless steel
- Feature a protective coating for extra durability and protection from rust and corrosion
- Satin-matte finish reduces eye-fatiguing glare
Spécifications
- Tip Configuration:Serrated tips
- Overall Length:305 mm