$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
E. COLI O 55: K 59 ANTISERA 3ML FOR IHC
75785-632
:
PRO-LAB DIAGNOSTICS INC CA
E. COLI O 55: K 59 ANTISERA 3ML FOR IHC
75785-632
:
PRO-LAB DIAGNOSTICS INC CA
:
PLA525
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, PP
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International