Microbank Cryoblock, Pro-Lab Diagnostics
Microbank Cryoblock, Pro-Lab Diagnostics
Microbank™ Cryo Cube Base
Catalog # 75784-934
Supplier #:  PL.156
Microbank Cryoblock, Pro-Lab Diagnostics
Catalog # 75784-934
Microbank Cryoblock, Pro-Lab Diagnostics
Microbank Cryoblock, Pro-Lab Diagnostics
Microbank™ Cryo Cube Base
Price
$175.10
Log in for availability
Pack of 4
Product Details & Documents
A 20 well, insulated cryoblock is available which enables the operator to remove a number of Microbank™ vials from the freezer for sub culture without risk of the culture beads defrosting.
The cryoblock is insulated with a polystyrene shield.
Specifications
  • Désignation:
    Microbank™ Cryo Cube Base