Microbank Cryoblock, Pro-Lab Diagnostics | Avantor
Microbank Cryoblock, Pro-Lab Diagnostics
Microbank™ Cryoblock
Catalog # 75784-932
Fournisseur #:  PL.155-1
Microbank Cryoblock, Pro-Lab Diagnostics
Catalog # 75784-932
Microbank Cryoblock, Pro-Lab Diagnostics
Microbank Cryoblock, Pro-Lab Diagnostics
Microbank™ Cryoblock
Price
$565.02
Log in pour la disponibilité
Each
Product Details & Documents
A 20 well, insulated cryoblock is available which enables the operator to remove a number of Microbank™ vials from the freezer for sub culture without risk of the culture beads defrosting.
The cryoblock is insulated with a polystyrene shield.
Spécifications
  • Désignation:
    Microbank™ Cryoblock