Microbank™-Dry, Pro-Lab Diagnostics | Avantor
Microbank™-Dry, Pro-Lab Diagnostics
Microbank, dry, Assorted, EA
Catalog # 75784-892
Fournisseur #:  PL.172-M
Microbank™-Dry, Pro-Lab Diagnostics
Catalog # 75784-892
Microbank™-Dry, Pro-Lab Diagnostics
Microbank™-Dry, Pro-Lab Diagnostics
Microbank, dry, Assorted, EA
Price
$636.94
Log in pour la disponibilité
Pack of 80 (2ml)
Product Details & Documents
A ready-to-use system designed to simplify the storage and retrieval of bacterial and fungal cultures.
  • Longer survival of fastidious cultures
  • Higher quantitative recoveries
  • Utilizes porous glass beads and a specially formulated cryopreservative for storage at low temperatures
Each Microbank™ vial contains approximately 25 sterile coloured beads (single colour) and the cryopreservative
The long term storage of microorganisms is a significant challenge in microbiology. Microbank™ offers a platform that utilizes porous glass beads and a specially formulated cryopreservative for storage at low temperatures. This format coupled with the Microbank™ Cryoblock offers the least possibility of disturbance to the stored organism, yet, permits ready and rapid access.
Each Microbank™-Dry vial contains approximately 25 sterile coloured beads (single colour). The specially treated beads are of a porous nature allowing microorganisms to readily adhere onto the bead surface. After
the addition of cryopreservative and inoculation the Microbank™-Dry vials are kept at -70°C for extended storage. When a fresh culture is required, a single bead is easily removed from the Microbank™-Dry vial and used to directly inoculate a suitable culture medium.
Spécifications
  • Color:
    Assorted
  • Désignation:
    Microbank, dry
  • Packaging:
    EA