Density Sets | Avantor
Density Sets
Density Set, Polyvinyl Chloride (PVC)
Catalog # 470221-108
Supplier:   Wards
Fournisseur #:  PH0113G
Density Sets
Catalog # 470221-108
Supplier:  Wards
Density Sets
Density Sets
Density Set, Polyvinyl Chloride (PVC)
Price
$103.40
Log in pour la disponibilité
Each
Product Details & Documents
Students Learn That the Density of a Substance is Constant, Even as Its Mass and Volume Change.
  • Each set has 12 cylindrical bars
  • Bars are same diameter but different lengths
  • Each set comes in an attractive wooden holder
Spécifications
  • Désignation:
    Density Set, Polyvinyl Chloride (PVC)