Cap Inserts for Cryogenic Vials, Globe Scientific | Avantor
Cap Inserts for Cryogenic Vials, Globe Scientific
Cap inserts for cryogenic vials, Orange
Catalog # 75784-232
Supplier:   Globe Scientific
Fournisseur #:  30079N
Cap Inserts for Cryogenic Vials, Globe Scientific
Catalog # 75784-232
Supplier:  Globe Scientific
Cap Inserts for Cryogenic Vials, Globe Scientific
Cap Inserts for Cryogenic Vials, Globe Scientific
Cap inserts for cryogenic vials, Orange
Price
$120.26
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 1,000
Product Details & Documents
Produced from polypropylene, cap inserts are available in assorted colors and are ideal for color coding and sample identification
  • Polypropylene (PP)
  • Designed for use with globe scientific CryoClear cryogenic vials
  • Suitable for use on both internal and extenrally threaded caps
The inserts nest easily into place on both internal and external caps.
Ordering information: Cap inserts are packaged 1000 per bag.
Spécifications
  • Color:
    Orange
  • Désignation:
    Cap inserts for cryogenic vials