Cap Inserts for Cryogenic Vials, Globe Scientific
Cap inserts for cryogenic vials, Orange
Catalog # 75784-232
Supplier: Globe Scientific
Fournisseur #: 30079N
Jump to:
Product Details & Documents
Produced from polypropylene, cap inserts are available in assorted colors and are ideal for color coding and sample identification
- Polypropylene (PP)
- Designed for use with globe scientific CryoClear cryogenic vials
- Suitable for use on both internal and extenrally threaded caps
The inserts nest easily into place on both internal and external caps.
Ordering information: Cap inserts are packaged 1000 per bag.
Spécifications
- Color:Orange
- Désignation:Cap inserts for cryogenic vials