Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Adjustable Sampling Tubes for Bioreactors, M10 | Avantor

Adjustable Sampling Tubes for Bioreactors, M10

320 mm, 15 L, Adjustable sampling tube
Catalog # 75780-150
Supplier:   Chemglass
Fournisseur #:  CLS-1385-13
Adjustable Sampling Tubes for Bioreactors, M10
Catalog # 75780-150
Supplier:  Chemglass
Adjustable Sampling Tubes for Bioreactors, M10
Adjustable Sampling Tubes for Bioreactors, M10

320 mm, 15 L, Adjustable sampling tube

Price
$936.66
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

These sampling tubes are adjustable to permit the culture fluid to be sampled at any desired level.

Tubing is ¹/₄" OD. Stainless steel components manufactured from 316 L with an electropolished surface finish of 20Ra or better.

Spécifications
  • Volume:
    15 L
  • Désignation:
    Adjustable sampling tube
  • Longueur:
    320 mm