Staticide® Concentrate, ACL Staticide | Avantor
Staticide® Concentrate, ACL Staticide
Quart, 946.3 mL (32 oz.)
Catalog # 75780-008
Supplier:   ACL Staticide
Fournisseur #:  3000Q
Staticide® Concentrate, ACL Staticide
Catalog # 75780-008
Supplier:  ACL Staticide
Staticide® Concentrate, ACL Staticide
Staticide® Concentrate, ACL Staticide
Quart, 946.3 mL (32 oz.)
Price
$345.86
Log in pour la disponibilité
Each (1qt US)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Staticide® Concentrate is available to save freight, handling, and storage costs while offering greater flexibility in dilution.
Staticide® Concentrate can be diluted with tap waater, Isopropyl Alcohol, Carbon Tetrachloride, or Perchloroethylene to meet various processing needs.
Staticide® Concentrate should be mixed with water or other solvents in the following concentrate solvent ratios to obtain these formulations: General Purpose Staticide® 1:99, Heavy Duty Staticide® 1:39
 : When properly applied, Staticide® complies with or exceeds all electrostatic decay criteria established for antistatic performance as set forth by NFPA and MIL B-81705, when tested in accordance with Federal Test Std 101B, Method 4046.
 : Staticide® Concentrate is available in quart, gallon, and 50 gallon drum containers.
Spécifications
  • Désignation:
    Quart
  • Taille:
    946.3 mL (32 oz.)