TWD Barcoded Autosampler Vials
Cap only, black
Catalog # 75780-320
Jump to:
Discontinued
Product Details & Documents
TWD Barcoded Autosampler Vials with 2D barcode.
- Vials feature writing patch and graduations
- Glass construction
- Screw-top 9-425 GPI thread finish
TWD barcoded autosampler kits are a cost-effective option if you're looking for a easy, hassle-free option to track your samples your samples.
Delivery information: Convenient Kit includes black PP cap and red rubber/PTFE Septa; available as vial and cap kit or cap only.
Spécifications
- Désignation:Cap only, black