TWD Barcoded Autosampler Vials
Vial and cap kit, 2.00 ml, 12×32 mm, Glass, Amber, Oui, Red rubber/PTFE
Catalog # 75780-316
Supplier: TWD Scientific
Supplier #: GV1232A-P-2DK
Jump to:
Product Details & Documents
TWD Barcoded Autosampler Vials with 2D barcode.
- Vials feature writing patch and graduations
- Glass construction
- Screw-top 9-425 GPI thread finish
TWD barcoded autosampler kits are a cost-effective option if you're looking for a easy, hassle-free option to track your samples your samples.
Delivery information: Convenient Kit includes black PP cap and red rubber/PTFE Septa; available as vial and cap kit or cap only.
Specifications
- Avec bouchon:Oui
- Septum:Red rubber/PTFE
- Capacité:2.00 ml
- Color:Amber
- Désignation:Vial and cap kit
- Matière:Glass
- Diameter ×H:12×32 mm