Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Biox Cardboard Freezer Boxes

100, 5 cm (2")
  75780-314
 :  BX90-2200
Biox Cardboard Freezer Boxes
  75780-314
Biox Cardboard Freezer Boxes
Biox Cardboard Freezer Boxes

100, 5 cm (2")

 
$553.75
   
Case of 100
 

Premium cardboard freezer boxes with a water-resistant coating.

  • Chemically resistant to alcohols and mild organic solvents
  • Temperature range: –196 to 121°C (–320 to 250 °F)
  • Accommodates 0.5 ml, 1.5 ml, or 2.0 ml microtubes
  • Suitable for liquid nitrogen storage

Boxes feature a water-resistant coating for improved durability.

 
  • Nb de places:
    100
  • Hauteur:
    5 cm (2")