Polypropylene Vials, Starplex®
Unlabeled, 10.00 ml, Conical bottom, Clear, Non, None, —, Non sterile
Catalog # CA10861-530
Supplier: Starplex
: V8094
Jump to:
Product Details & Documents
These vials are made of crack-resistant medical grade polypropylene.
- With or without screw caps
- Flat, conical or round bottoms
Available clear for easy visual inspection and amber for light-sensitive components. These vials provide convenient storage for samples.
Spécifications
- Avec bouchon:Non
- Cap Type:None
- Sterility:Non sterile
- Capacité:10.00 ml
- Color:Clear
- Désignation:Unlabeled
- Cap Color:—
- Bottom Style:Conical bottom