Protocol Station, Restek | Avantor
Protocol Station, Restek
Protocol Station*, Brass, chrome plated, CGA 350 (H2, P5)
Catalog # 10854-418
Supplier:   Restek
Fournisseur #:  21348
Protocol Station, Restek
Catalog # 10854-418
Supplier:  Restek
Protocol Station, Restek
Protocol Station, Restek
Protocol Station*, Brass, chrome plated, CGA 350 (H2, P5)
Price
$1,373.98
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
The protocol station is designed for convenient wall mounting of high-purity gas regulators.
  • Fabricated in stainless steel or chrome-plated brass
  • Provides ease of use for high-purity gas regulators
Wall mounting provides ease of use, prevents gas regulator damage, and improves safety. Either chrome-plated brass or 316 stainless steel option is complete with a 3-foot, flexible, all-stainless-steel pigtail with armor casing. The CGA connection on the pigtail has an integral check valve in the gland to prevent contamination during cylinder changeout.
Ordering information: Pressure regulator not included. Order separately.
Spécifications
  • Désignation:
    Protocol Station*
  • Matière:
    Brass, chrome plated
  • Includes:
    CGA 350 (H2, P5)
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.