Protocol Station, Restek
Protocol Station*, Brass, chrome plated, CGA 580 (N2, He, Ar)
  10854-412
 :   Restek
 :  21347
Protocol Station, Restek
  10854-412
 :  Restek
Protocol Station, Restek
Protocol Station, Restek
Protocol Station*, Brass, chrome plated, CGA 580 (N2, He, Ar)
$1,387.62
Each
The protocol station is designed for convenient wall mounting of high-purity gas regulators.
  • Fabricated in stainless steel or chrome-plated brass
  • Provides ease of use for high-purity gas regulators
Wall mounting provides ease of use, prevents gas regulator damage, and improves safety. Either chrome-plated brass or 316 stainless steel option is complete with a 3-foot, flexible, all-stainless-steel pigtail with armor casing. The CGA connection on the pigtail has an integral check valve in the gland to prevent contamination during cylinder changeout.
 : Pressure regulator not included. Order separately.
  • Désignation:
    Protocol Station*
  • Matière:
    Brass, chrome plated
  • Includes:
    CGA 580 (N2, He, Ar)