Inserts for Versa Vials™, Restek
Versa vial insert, 0.25 ml, Flat bottom, Glass, Non
Catalog # 10854-370
Supplier: Restek
Fournisseur #: 22712
Jump to:
Product Details & Documents
Glass and polypropylene Versa Vial™ inserts are compatible with Versa Vials™ of various sizes.
- 250 µl volume vial compatibility
- Glass or polypropylene construction
Inserts for Versa Vials™ are made of rugged polypropylene or glass and feature a flat-bottomed design or flanges.
Spécifications
- Avec bouchon:Non
- Capacité:0.25 ml
- Désignation:Versa vial insert
- Matière:Glass
- Bottom Style:Flat bottom