Inserts for Versa Vials™, Restek
Versa vial insert with flange, 0.25 ml, Conical bottom, PP, Non
10854-364
: Avantor
Glass and polypropylene Versa Vial™ inserts are compatible with Versa Vials™ of various sizes.
- 250 µl volume vial compatibility
- Glass or polypropylene construction
Inserts for Versa Vials™ are made of rugged polypropylene or glass and feature a flat-bottomed design or flanges.
- Avec bouchon:Non
- Capacité:0.25 ml
- Désignation:Versa vial insert with flange
- Matière:PP
- Bottom Style:Conical bottom