Inserts for Versa Vials™, Restek
Versa vial insert with flange, 0.25 ml, Conical bottom, PP, Non
  10854-364
 :   Avantor
Inserts for Versa Vials™, Restek
  10854-364
 :  Avantor
Inserts for Versa Vials™, Restek
Inserts for Versa Vials™, Restek
Versa vial insert with flange, 0.25 ml, Conical bottom, PP, Non
$26.24
Pack of 100
Glass and polypropylene Versa Vial™ inserts are compatible with Versa Vials™ of various sizes.
  • 250 µl volume vial compatibility
  • Glass or polypropylene construction
Inserts for Versa Vials™ are made of rugged polypropylene or glass and feature a flat-bottomed design or flanges.
  • Avec bouchon:
    Non
  • Capacité:
    0.25 ml
  • Désignation:
    Versa vial insert with flange
  • Matière:
    PP
  • Bottom Style:
    Conical bottom