Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek

Insert for WISP 48 vials, 0.50 ml, Flat bottom, Glass, Non
  10854-094
 :   Restek
 :  21787
Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek
  10854-094
 :  Restek
Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek
Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek

Insert for WISP 48 vials, 0.50 ml, Flat bottom, Glass, Non

 
$59.22
 
   
Pack of 100
 

A variety of inserts compatible with screw-thread and crimp-top vials are available in a flat-bottomed format.

  • Screw-thread and crimp-top vial compatibility
  • Can also be used with snap seal vials
  • Polypropylene construction

Inserts for screw-thread, crimp-top, and snap seal vials are made of rugged polypropylene and feature a flat-bottomed design.

 : Polypropylene inserts available on request (1000-packs only). Also fit 4.0 ml Snap Seal vials.

 
  • Avec bouchon:
    Non
  • Capacité:
    0.50 ml
  • Désignation:
    Insert for WISP 48 vials
  • Matière:
    Glass
  • Bottom Style:
    Flat bottom