Well Plates, Restek
Well Plates, Restek
1.3 mL 96-Well Plates, Round, Round
Catalog # 10853-450
Supplier:   Restek
Supplier #:  26495
Well Plates, Restek
Catalog # 10853-450
Supplier:  Restek
Well Plates, Restek
Well Plates, Restek
1.3 mL 96-Well Plates, Round, Round
Price
$59.85
New Customer Promotional Price
Log in for availability
Pack of 5
Product Details & Documents
Polypropylene plates with round-bottom wells reduce liquid retention; conical bottom provides optimal recovery of reagents.
  • Round well shape is ideal for applications that require vortexing
  • Ideal for sample collection, storage, sampling, and combinatorial chemistry and library applications
  • Fit most autosampler compartments
Nunc® shared wall technology allows increased well volume for optimum storage capacity and improved mixing.
Certifications: All microplates manufactured by Nunc® meet the recommendation of American National Standards Institute (ANSI) (ANSI/SBS 1-2004).
Specifications
  • Désignation:
    1.3 mL 96-Well Plates
  • Well Shape:
    Round
  • Well Bottom Shape:
    Round