Well Plates, Restek
1.3 mL 96-Well Plates, Round, Round
Catalog # 10853-450
Supplier: Restek
Supplier #: 26495
Jump to:
Product Details & Documents
Polypropylene plates with round-bottom wells reduce liquid retention; conical bottom provides optimal recovery of reagents.
- Round well shape is ideal for applications that require vortexing
- Ideal for sample collection, storage, sampling, and combinatorial chemistry and library applications
- Fit most autosampler compartments
Nunc® shared wall technology allows increased well volume for optimum storage capacity and improved mixing.
Certifications: All microplates manufactured by Nunc® meet the recommendation of American National Standards Institute (ANSI) (ANSI/SBS 1-2004).
Specifications
- Désignation:1.3 mL 96-Well Plates
- Well Shape:Round
- Well Bottom Shape:Round