Well Plates, Restek
1.3 mL 96-Well Plates, Round, Round
  10853-266
 :   Restek
 :  26494
Well Plates, Restek
  10853-266
 :  Restek
Well Plates, Restek
Well Plates, Restek
1.3 mL 96-Well Plates, Round, Round
$462.28
Pack of 50
Polypropylene plates with round-bottom wells reduce liquid retention; conical bottom provides optimal recovery of reagents.
  • Round well shape is ideal for applications that require vortexing
  • Ideal for sample collection, storage, sampling, and combinatorial chemistry and library applications
  • Fit most autosampler compartments
Nunc® shared wall technology allows increased well volume for optimum storage capacity and improved mixing.
 : All microplates manufactured by Nunc® meet the recommendation of American National Standards Institute (ANSI) (ANSI/SBS 1-2004).
  • Désignation:
    1.3 mL 96-Well Plates
  • Well Shape:
    Round
  • Well Bottom Shape:
    Round