Lead Shields
Lead Shields for 1 - 20 Disk Sources
Catalog # 470220-460
Supplier: SPECTRUM TECHNIQUES, LLC
Fournisseur #: LSD5
Jump to:
Product Details & Documents
Lead shield for shipping and storage.
- Easy solution for storage
- Keep materials and students safe
Lead Shield simplifies shipping problems and offers ultra-safe storage of sources. Different options based on the numbers of samples you need to store.
Spécifications
- Désignation:Lead Shields for 1 - 20 Disk Sources