High-Temperature String, Restek
High-Temperature String
Catalog # 10852-880
Supplier: Restek
Supplier #: 20618
Jump to:
Product Details & Documents
Use high-temperature string to restring capillary columns, attach column connectors to column cages, or hold the column in the GC oven.
Capable of withstanding temperatures to 400 °C.
Specifications
- Désignation:High-Temperature String