High-Temperature String, Restek
High-Temperature String, Restek
High-Temperature String
Catalog # 10852-880
Supplier:   Restek
Supplier #:  20618
High-Temperature String, Restek
Catalog # 10852-880
Supplier:  Restek
High-Temperature String, Restek
High-Temperature String, Restek
High-Temperature String
Price
$364.46
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Use high-temperature string to restring capillary columns, attach column connectors to column cages, or hold the column in the GC oven.
Capable of withstanding temperatures to 400 °C.
Specifications
  • Désignation:
    High-Temperature String