Rethreading Tool, Restek | Avantor
Rethreading Tool, Restek
Rethreading Tool
Catalog # 10852-866
Supplier:   Restek
Fournisseur #:  23017
Rethreading Tool, Restek
Catalog # 10852-866
Supplier:  Restek
Rethreading Tool, Restek
Rethreading Tool, Restek
Rethreading Tool
Price
$295.85
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Rethreading tools repair worn or damaged threads that come about as a result of consistent use and temperature changes.
  • Multiple uses (fittings, injectors, detectors, etc.)
  • Built-in guide to prevent cross-threading
Repeated installation, removal, and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.
Spécifications
  • Désignation:
    Rethreading Tool
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.