Rethreading Tool, Restek
Rethreading Tool
Catalog # 10852-864
Supplier: Restek
Fournisseur #: 23016
Jump to:
Product Details & Documents
Rethreading tools repair worn or damaged threads that come about as a result of consistent use and temperature changes.
- Multiple uses (fittings, injectors, detectors, etc.)
- Built-in guide to prevent cross-threading
Repeated installation, removal, and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.
Spécifications
- Désignation:Rethreading Tool