Rethreading Tool, Restek | Avantor
Rethreading Tool, Restek
Rethreading Tool
Catalog # 10852-864
Supplier:   Restek
Fournisseur #:  23016
Rethreading Tool, Restek
Catalog # 10852-864
Supplier:  Restek
Rethreading Tool, Restek
Rethreading Tool, Restek
Rethreading Tool
Price
$197.97
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Rethreading tools repair worn or damaged threads that come about as a result of consistent use and temperature changes.
  • Multiple uses (fittings, injectors, detectors, etc.)
  • Built-in guide to prevent cross-threading
Repeated installation, removal, and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.
Spécifications
  • Désignation:
    Rethreading Tool