Rethreading Tool, Restek | Avantor
Rethreading Tool, Restek
Rethreading Tool
Catalog # 10852-856
Rethreading Tool, Restek
Catalog # 10852-856
Supplier:  
Rethreading Tool, Restek
Rethreading Tool, Restek
Rethreading Tool
Discontinued
Product Details & Documents
Rethreading tools repair worn or damaged threads that come about as a result of consistent use and temperature changes.
  • Multiple uses (fittings, injectors, detectors, etc.)
  • Built-in guide to prevent cross-threading
Repeated installation, removal, and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.
Spécifications
  • Désignation:
    Rethreading Tool