Rtx®-5 GC Columns, Restek
Rtx-5 Agilent 6850 configuration, Capillaire, 0.50 µm, 0.25 mm, 30 m
10849-666
A low-polarity general-purpose column for the analysis of pesticides, PCBs, aromatic hydrocarbons, drugs, and essential oils.
- Extremely low bleed
- Low-polarity column
- Internal dimensions and film thickness vary between models
Equivalent to USP G27, G36 phases. All residual catalysts and low molecular weight fragments are removed from the Rtx®-5 polymer, providing a tight mono-modal distribution and extremely low bleed. Phase composition: Crossbond® 5% diphenyl, 95% dimethylpolysiloxane.
- Type de colonne:Capillaire
- Épaisseur du film:0.50 µm
- Ø int.:0.25 mm
- Désignation:Rtx-5 Agilent 6850 configuration
- Longueur:30 m
- Cat. No.:10849-666
- Temperature limit:–60...350 °C
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



