Ionizing Nozzles, Transforming Technologies | Avantor
Ionizing Nozzles, Transforming Technologies
Manifold short (4 - 12") nozzle tip
Catalog # 10837-078
Ionizing Nozzles, Transforming Technologies
Catalog # 10837-078
Supplier:  
Ionizing Nozzles, Transforming Technologies
Ionizing Nozzles, Transforming Technologies
Manifold short (4 - 12") nozzle tip
Discontinued
Product Details & Documents
Transforming Technologies’ Ionizing Nozzles can handle unique and difficult point-of-use ionization applications with interchangeable nozzle tips
Ionizing Nozzles are compatible with all IN Gun series ionizers.
Ionizing nozzles can be mounted in hard to reach areas that larger ionizers simply cannot handle. Nozzle tips are designed to spread ionized air in many unique ways. Wide angle, long range, bendable, luer lock, or manifold tips may be substituted for the N0010 tip, depending upon the application. (N0010 is the standard tip sold with Transforming Technologies Ionizing Nozzles).
Spécifications
  • Désignation:
    Manifold short (4 - 12") nozzle tip
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.