Mounting Medium, Azer Scientific
Mounting Medium, Azer Scientific
S-Mounting Medium, 16 oz.
Catalog # CA10821-474
Supplier:   Azer Scientific
Supplier #:  ES724
Mounting Medium, Azer Scientific
Catalog # CA10821-474
Supplier:  Azer Scientific
CAS Number:  
Mounting Medium, Azer Scientific
Mounting Medium, Azer Scientific
S-Mounting Medium, 16 oz.
CAS Number:  
Price
$2,961.96
Log in for availability
Case of 12 (1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Adhesive mounting medium with permanent clarity for long term slide storage.
  • Traditional formula or quick drying acrylic
  • Low viscosity for easy cover glass mounting
Non-aqueous S-mounting medium formula is compatible with xylene and most xylene substitutes, dries quickly and and will not crack or yellow over time.
Traditional mounting medium is soluble in xylene and toluene, dries optically clear and will resist photobleaching and stain fading.
Specifications
Specifications
  • Désignation:
    S-Mounting Medium
  • Taille:
    16 oz.