Cabinet Accessories, Justrite®
Lithium Label for Hazardous Cabinets, Any Hazardous Material cabinet, 12", 50", 0.5 lb.
Catalog # 10836-878
Supplier: Justrite
Fournisseur #: 29018
Jump to:
Product Details & Documents
Cabinet accessories offer an opportunity to expand storage and add new convenience to existing cabinets, or customize new cabinets with additional shelves to meet specific workflow needs.
- Expand storage
- Add new convenience
- Customize new cabinets
Polyethylene trays offer additional protection and have a slightly raised curved edge to contain spills
SpillSlope™ galvanized steel shelves hold up to 159 kg (350 lbs.) and adjust on 5.7 cm (21/4") increments for versatile storage.
They may also be used in the bottom sill area to catch incidental spills and leaks.
Spécifications
- Largeur:12"
- Désignation:Lithium Label for Hazardous Cabinets
- Hauteur:50"
- Shipping Weight:0.5 lb.
- For Use With:Any Hazardous Material cabinet



