Cabinet Accessories, Justrite®
Label Pack for Hazardous Cabinets, Any Hazardous Material cabinet, 10", 69", 0.5 lb.
Catalog # 10836-876
Supplier: Justrite
Supplier #: 29017
Jump to:
Product Details & Documents
Cabinet accessories offer an opportunity to expand storage and add new convenience to existing cabinets, or customize new cabinets with additional shelves to meet specific workflow needs.
- Expand storage
- Add new convenience
- Customize new cabinets
Polyethylene trays offer additional protection and have a slightly raised curved edge to contain spills
SpillSlope™ galvanized steel shelves hold up to 159 kg (350 lbs.) and adjust on 5.7 cm (21/4") increments for versatile storage.
They may also be used in the bottom sill area to catch incidental spills and leaks.
Spécifications
- Largeur:10"
- Désignation:Label Pack for Hazardous Cabinets
- Hauteur:69"
- Shipping Weight:0.5 lb.
- For Use With:Any Hazardous Material cabinet



