Ionizing Nozzles, Transforming Technologies | Avantor
Ionizing Nozzles, Transforming Technologies
Bendable 16" nozzle tip
Catalog # 10837-070
Fournisseur #:  N0050-16
Ionizing Nozzles, Transforming Technologies
Catalog # 10837-070
Ionizing Nozzles, Transforming Technologies
Ionizing Nozzles, Transforming Technologies
Bendable 16" nozzle tip
Price
$146.00
Log in pour la disponibilité
Each
Product Details & Documents
Transforming Technologies’ Ionizing Nozzles can handle unique and difficult point-of-use ionization applications with interchangeable nozzle tips
Ionizing Nozzles are compatible with all IN Gun series ionizers.
Ionizing nozzles can be mounted in hard to reach areas that larger ionizers simply cannot handle. Nozzle tips are designed to spread ionized air in many unique ways. Wide angle, long range, bendable, luer lock, or manifold tips may be substituted for the N0010 tip, depending upon the application. (N0010 is the standard tip sold with Transforming Technologies Ionizing Nozzles).
Spécifications
  • Désignation:
    Bendable 16" nozzle tip