Ionizing Nozzles, Transforming Technologies
Ionizing Nozzles, Transforming Technologies
Long range nozzle tip
Catalog # 10837-066
Supplier #:  N0040
Ionizing Nozzles, Transforming Technologies
Catalog # 10837-066
Ionizing Nozzles, Transforming Technologies
Ionizing Nozzles, Transforming Technologies
Long range nozzle tip
Price
$73.00
Log in for availability
Each
Product Details & Documents
Transforming Technologies’ Ionizing Nozzles can handle unique and difficult point-of-use ionization applications with interchangeable nozzle tips
Ionizing Nozzles are compatible with all IN Gun series ionizers.
Ionizing nozzles can be mounted in hard to reach areas that larger ionizers simply cannot handle. Nozzle tips are designed to spread ionized air in many unique ways. Wide angle, long range, bendable, luer lock, or manifold tips may be substituted for the N0010 tip, depending upon the application. (N0010 is the standard tip sold with Transforming Technologies Ionizing Nozzles).
Specifications
  • Désignation:
    Long range nozzle tip