Polyethylene Plug Caps, Starplex®
Plug Style Caps, Dark Blue, Non sterile
Catalog # 10836-536
Fournisseur #: V9222
Jump to:
Discontinued
Product Details & Documents
Nonsterile polyethylene plug style caps are designed for use with 5mL and 10mL vials
All caps are bulk packaged in bags.
Spécifications
- Sterility:Non sterile
- Désignation:Plug Style Caps
- Cap Color:Dark Blue
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



