Polyethylene Plug Caps, Starplex®
Plug Style Caps, Yellow, Non sterile
Catalog # 10836-534
Fournisseur #: V9221
Jump to:
Discontinued
Product Details & Documents
Nonsterile polyethylene plug style caps are designed for use with 5mL and 10mL vials
All caps are bulk packaged in bags.
Spécifications
- Sterility:Non sterile
- Désignation:Plug Style Caps
- Cap Color:Yellow