Polyethylene Plug Caps, Starplex®
Plug Style Caps, White, Non sterile
Catalog # 10836-532
: V9220
Jump to:
Discontinued
Product Details & Documents
Nonsterile polyethylene plug style caps are designed for use with 5mL and 10mL vials
All caps are bulk packaged in bags.
Spécifications
- Sterility:Non sterile
- Désignation:Plug Style Caps
- Cap Color:White