Polyethylene Plug Caps, Starplex® | Avantor
Polyethylene Plug Caps, Starplex®
Plug Style Caps, Red, Non sterile
Catalog # 10836-530
Fournisseur #:  V9200
Polyethylene Plug Caps, Starplex®
Catalog # 10836-530
Supplier:  
Polyethylene Plug Caps, Starplex®
Plug Style Caps, Red, Non sterile
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
PLUG CAPS ONLY RED CS6000 Catalog #CA11007-698
Product Details & Documents
Nonsterile polyethylene plug style caps are designed for use with 5mL and 10mL vials
All caps are bulk packaged in bags.
Spécifications
  • Sterility:
    Non sterile
  • Désignation:
    Plug Style Caps
  • Cap Color:
    Red