Stoppers, Penny-Head, Glass, Solid, Chemscience
Stoppers, Penny-Head, Glass, Solid, Chemscience
Stopper, penny-head, glass, solid, 10/18
Catalog # CA10823-176
Supplier:   CHEM SCIENCE INC CA
Supplier #:  CS-S0291018
Stoppers, Penny-Head, Glass, Solid, Chemscience
Catalog # CA10823-176
Stoppers, Penny-Head, Glass, Solid, Chemscience
Stoppers, Penny-Head, Glass, Solid, Chemscience
Stopper, penny-head, glass, solid, 10/18
Price
$159.43
Log in pour la disponibilité
Case of 10
Product Details & Documents
Solid penny head glass stoppers are with an interchangeable ground joint.
Spécifications
  • ST Joint No.:
    10/18
  • Désignation:
    Stopper, penny-head, glass, solid