Individual Slotted Killogram Weights
Slotted Weight, 5 kg
Catalog # 470219-780
Supplier: Wards
Fournisseur #: PH0264B4
Jump to:
Product Details & Documents
Individual cast iron slotted weights.
- Available individually
- Stackable
- Made from solid cast iron
- Finished with a black enamel
- Stamped with weight value
Slotted iron masses are suitable for use with sonometers or other applications, where a large load is required. The slotted masses can be hung on a hanger.
Ordering information: Hanger ordered separately.
Spécifications
- Désignation:Slotted Weight
- Weight:5 kg
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Slotted Hanger Only, 0.5 kg