Individual Slotted Killogram Weights
Slotted Weight, 1 kg
Catalog # 470219-776
Supplier:   Wards
 :  PH0264B2
Individual Slotted Killogram Weights
Catalog # 470219-776
Supplier:  Wards
Individual Slotted Killogram Weights
Individual Slotted Killogram Weights
Slotted Weight, 1 kg
Price
$40.90
Log in pour la disponibilité
Each
Product Details & Documents
Individual cast iron slotted weights.
  • Available individually
  • Stackable
  • Made from solid cast iron
  • Finished with a black enamel
  • Stamped with weight value
Slotted iron masses are suitable for use with sonometers or other applications, where a large load is required. The slotted masses can be hung on a hanger.
 : Hanger ordered separately.
Spécifications
  • Désignation:
    Slotted Weight
  • Weight:
    1 kg
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Slotted Hanger Only, 0.5 kg
    Slotted Hanger Only, 0.5 kg
    470229-366
    Log in pour la disponibilité
    Each - $37.90