Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Whatman™ Polyester Drain Discs | Avantor

Whatman™ Polyester Drain Discs

Polyester drain disc, PE, membrane filter accessories, 10 mm, 100 µm
Catalog # CA15000-602
Supplier:   Cytiva
Fournisseur #:  230300
Whatman™ Polyester Drain Discs
Catalog # CA15000-602
Supplier:  Cytiva
Whatman™ Polyester Drain Discs
Whatman™ Polyester Drain Discs

Polyester drain disc, PE, membrane filter accessories, 10 mm, 100 µm

Price
$110.60
Log in pour la disponibilité
Pack of 100
Product Details & Documents

The polyester drain disc is binder free and has a thickness of 100 µm.

  • Flat surface to eliminate filter tearing or rupturing
  • Used as a separator between membrane layers in serial stack filtration applications

This chemically inert support disc is available in a variety of diameters for use in a range of devices. With Application in General laboratory microfiltration, Quality control and sterility testing, Removal of particulates from HPLC solvents, Tissue culture media filtration.

Spécifications
  • Ø:
    10 mm
  • Désignation:
    Polyester drain disc, PE, membrane filter accessories
  • Épaisseur:
    100 µm
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.