Eudiometer
Eudiometer, 50 ml
Catalog # 470219-674
Supplier: VWR International
Jump to:
Product Details & Documents
Demonstrate the combining volume ratios of gases.
- Two sizes available
- Platinum electrodes included
Long, sturdy glass tubes have platinum electrodes sealed at the closed end with a 2 mm arc gap. The tubes are marked with a permanent brown scale in 0.1 or 0.2 mL divisions for 50 and 100 mL respectively. An induction coil is recommended for use with eudiometers.
Spécifications
- Désignation:Eudiometer, 50 ml