Water Accelerometer
Product Details & Documents
Visualize acceleration and centripetal force with this unique device.
- Dramatic visual demonstration of accelerated vs equilibrium motion
- Liquid surface shows relative acceleration of a moving system
- Printed grid allows for quntitative evaluation of slope
- Turntable provides easy method for studying rotating systems
- Student lab sheet provided
The unit is comprised of a clear rectangular container, marked with a centimeter grid, and partially filled with water. At equilibrium, the water forms a level, horizontal line on the grid. When the device is accelerated, the slope of the water level changes, allowing a quantitative analysis of Newton’s 2nd Law. Affix to a dynamics cart to compare acceleration down an inclined plane at different angles. Or, visualize the acceleration of a half-atwood machine under different conditions. The unit can also be attached to a turntable, allowing students to study the centrifugal effects of rotation.
Ordering information: Accelerometer and turntable available separately. Unit shipped dry; water not included. Recommended: Use colored water to enhance the visual impact for a demonstration. Use a camera to accurately capture and evaluate the slope.
Spécifications
- Désignation:Water accelerometer only