VWR® Wall Shelf Brackets, Fixed and Adjustable
Book End Shelf Brackets, Light neutral, 18" shelf, 18"
Catalog # CFM-0062-AD
Supplier: VWR International
Jump to:
Product Details & Documents
Fixed and adjustable wall-mounted brackets.
- Designed to suit a wide array of storage applications
- Fixed brackets mount directly to the wall
- Adjustable brackets are designed to mount to vertical wall supports (sold separately)
Fixed brackets features a flat top surface for simple shelf attachment. Adjustable brackets features a lipped top surface to provide increased security and stability when shelves are attached.
Ordering information: Fixed shelf brackets are supplied without mounting hardware. Adjustable brackets come with mounting hardware.
Spécifications
- Profondeur:18"
- Color:Light neutral
- Désignation:Book End Shelf Brackets
- For Use With:18" shelf
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



