VWR® Wall Shelf Brackets, Fixed and Adjustable | Avantor
VWR® Wall Shelf Brackets, Fixed and Adjustable
Book End Shelf Brackets, 12" shelf, 12"
Catalog # CFM-0060-AD
Supplier:   VWR International
VWR® Wall Shelf Brackets, Fixed and Adjustable
Catalog # CFM-0060-AD
Supplier:  VWR International
VWR® Wall Shelf Brackets, Fixed and Adjustable
VWR® Wall Shelf Brackets, Fixed and Adjustable
Book End Shelf Brackets, 12" shelf, 12"
Price
$174.03
Log in pour la disponibilité
Each
Product Details & Documents
Fixed and adjustable wall-mounted brackets.
  • Designed to suit a wide array of storage applications
  • Fixed brackets mount directly to the wall
  • Adjustable brackets are designed to mount to vertical wall supports (sold separately)
Fixed brackets features a flat top surface for simple shelf attachment. Adjustable brackets features a lipped top surface to provide increased security and stability when shelves are attached.
Ordering information: Fixed shelf brackets are supplied without mounting hardware. Adjustable brackets come with mounting hardware.
Spécifications
  • Profondeur:
    12"
  • Désignation:
    Book End Shelf Brackets
  • For Use With:
    12" shelf